Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integra™ Miltex™ Mid Grade Hemostatic Forceps
Click to view available options
Tip Style:
Curved
Straight
Description
- Mid-grade
Specifications
Specifications
Tip Style | Straight |
Length (English) | 3.5 in. |
Length (Metric) | 89 mm |
Product Type | Forceps, Hemostatic Mosquito |
Quantity (No. per Case) | Each |
Grade | Mid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction