Learn More
3M™ Integrated Protective Eyewear
3M™ Integrated Protective Eyewear is designed with a tooless clip-on attachment to connect to the headband of the helmet or headgear. Easy to adjust and retracts neatly between the hard hat shell and suspension when not in use.
Supplier: 3M™ 7100205235
This item is not returnable.
View return policy
Description
- Attaches directly to safety helmet suspension
- Without temple arms, the helmet integrated safety glasses will not interfere with the acoustic seal of 3M helmet mounted earmuffs
- Retracts between safety helmet suspension and shell, helping to protect the lens when not in use
- New improved nose bridge design adds extra protection for the lenses, preventing scratches from the helmet
- Increased link friction provides even more secure positioning than the 3M™ Safety Glasses HIE6 Series
- Three pivot adjustment points allow easy maneuvering and adjustment for closeness and height
- Designed to be lightweight on nose and temples
- Anti-fog lens
Specifications
Amber | |
SecureFit™ Safety Helmet X5000 Series, H-700 Series, and H-800 Series of Hard Hats | |
20/Cs. | |
Polycarbonate |
Hard Hat Accessories | |
Sanding; Electrical; Facility Maintenance; Remediation; Grinding; Mining; Road construction; Oil and gas; Demolition; Construction; Machine operation; Heavy and light industry; Forestry | |
HIE603AF |