Description
- Attaches directly to safety helmet suspension
- Without temple arms, the helmet integrated safety glasses will not interfere with the acoustic seal of 3M helmet mounted earmuffs
- Retracts between safety helmet suspension and shell, helping to protect the lens when not in use
- New improved nose bridge design adds extra protection for the lenses, preventing scratches from the helmet
- Increased link friction provides even more secure positioning than the 3M™ Safety Glasses HIE6 Series
- Three pivot adjustment points allow easy maneuvering and adjustment for closeness and height
- Designed to be lightweight on nose and temples
- Anti-fog lens
Specifications
Specifications
| Lens Tint | Clear |
| Product Type | Hard Hat Accessories |
| For Use With (Equipment) | SecureFit™ Safety Helmet X5000 Series, H-700 Series, and H-800 Series of Hard Hats |
| For Use With (Application) | Sanding; Electrical; Facility Maintenance; Remediation; Grinding; Mining; Road construction; Oil and gas; Demolition; Construction; Machine operation; Heavy and light industry; Forestry |
| No. per Case | 20/Cs. |
| Model | V901 |
| Frame Material | Polycarbonate |