Learn More
3M™ Integrated Protective Eyewear
Catalog No. p-7182382 Shop All 3M Products
Click to view available options
Lens Tint:
Gray
This item is not returnable. View return policy
This item is not returnable. View return policy
3M™ Integrated Protective Eyewear is lightweight retractable eyewear that is easy to attach to the headband of 3M hard hats
  • Attaches directly to hard hat suspension, nearly eliminating the weight of the eyewear on the nose and temples
  • Retracts between hard hat suspension and shell, helping to protect the lens when not in use
  • Stable, stays in place and secure when not in use
  • Easy to maneuver and adjust for closeness and height
  • Comfort: No temples, minimal weight on nose bridge
  • Anti-fog lens helps prevent moisture from building up on the lens
  • Connects directly to the ratchet and pinlock suspension systems of the 3M™ H-700 and 3M™ H-800 Series of hard hats
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI Z89.1 2010
Lens Tint Gray
Product Type Integrated Protective Eyewear
For Use With (Equipment) 3M™ H 700 and H 800 Series of hard hats, 3M™ SecureFit™ Safety Helmet X5000
For Use With (Application) Assembly, Grinding, Machine Operations, Painting, Remediation, Sanding, Welding
No. per Case 20/Cs.
Model HIE603AF
Lens Material Polycarbonate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.