Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Integrin β1, Rat, Alexa Fluor 647, Clone: MB1.2,

Catalog No. MAB1997AF64 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Alexa Fluor 647
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
MAB1997AF64 100 μL Alexa Fluor 647
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1997AF64 Supplier MilliporeSigma™ Supplier No. MAB1997AF647
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Integrin beta-1 (UniProt P09055; also known as CD29, Fibronectin receptor subunit beta, GpIIa, VLA-1/2/3/4/5/6 subunit beta) is encoded by the Itgb1 (also known as Fnrb) gene (Gene ID 16412) in murine species. The integrin family of cell adhesion receptors consists of at least 16 membrane-associated heterodimers, composed of an alpha and beta subunit that associate in a non-covalent manner. The structure and functional diversity of the integrin family are based upon the pairing abilities of the individual alpha and beta subunits. Key to these molecular interactions between the integrin receptors and their respective ligands is the recognition of the Arg-Gly-Asp (RGD) sequence known to be present in the extracellular matrix components fibronectin, vitronectin, collagen, fibrinogen, and von Willebrand factor. Integrin beta-1 is a 130kDa transmembrane glycoprotein that interact with various integrin alpha subunits (including alpha 1, alpha 2, alpha 3, alpha 4, alpha 5, and alpha 6) to form the functional receptor complexes that bind to specific extracellular matrix proteins. Integrin receptors regulate a variety of important biological functions, including embryonic development, wound repair, hemostasis, and prevention of programmed cell death. They are also implicated in abnormal pathological states such as tumor directed angiogenesis, tumor growth and metastasis. These heterodimeric receptors bridge the cytoplasmic actin cytoskeleton with proteins present in the extracellular matrix and/or on adjacent cells. Interactions between integrins and the extracellular matrix lead to the activation of signal transduction pathways and regulation of gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β1
Applications Immunocytochemistry
Classification Monoclonal
Clone MB1.2
Concentration Please refer to lot specific datasheet.
Conjugate Alexa Fluor 647
Formulation Purified rat IgG2a Alexa Fluor™ 647 conjugate in buffer containing PBS, 1.5% BSA with 0.05% Sodium Azide.
Gene Itgb1, Fnrb
Gene Accession No. P09055
Host Species Rat
Immunogen Mouse Integrin beta-1 (Von Ballestrem, C.G., et al. (1996). Hybridoma. 15(2):125-132).
Purification Method Protein G Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics & Nuclear Function
Primary or Secondary Primary
Gene ID (Entrez) NP_034708
Test Specificity The specificity of MB1.2 was determined by its binding to antigen, and the ability of antiserum against the carboxyl terminal of VLA-beta 1 subunit to deplete antigens for MB1.2 in sequential immunoprecipitation experiments. MB1.2 had no effect on cell adhesion to matrix proteins (Von Ballestrem, C.G., et al. (1996). Hybridoma. 15(2):125-132).
Target Species Mouse
Content And Storage Stable for 1 year at 2°-8°C from date of receipt.
Isotype IgG2a κ
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.