Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin β7 Chain Rat anti-Mouse, Brilliant Violet 711, Clone: M293, BD Optibuild™

Catalog No. BDB743791 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB743791 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB743791 Supplier BD Biosciences Supplier No. 743791
Add to Cart
Add to Cart

Rat Monoclonal Antibody

The M293 monoclonal antibody specifically recognizes the integrin ß7 chain (also known as integrin ßp), which is found in association with either of twoαchains.α4ß7 (LPAM-1) is expressed on most mature lymphocytes and on small subsets of thymic and bone marrow cells; it interacts with several ligands, including VCAM-1, fibronectin, and MAdCAM-1.αIELß7 is found primarily on intestinal mucosal and intraepithelial lymphocytes (IEL), and it may be primarily involved in interactions between IEL and epithelia via /recognition of E-cadherin. The epitope recognized by the mAb M293 has been mapped to the region between amino acids 387 and 542. The M293 antibody exhibits little or no activity in blocking ß7 integrin-mediated adhesion.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 711nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV711 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 711 nm. This dye offers a very bright choice for the violet laser. Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. BV711 will also have moderate spillover into the BD Horizon Brilliant™ Violet 786 detector.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin β7 Chain
Classification Monoclonal
Clone M293
Concentration 0.2mg/mL
Conjugate Brilliant Violet 711
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Itgb7, Integrin βp, Integrin beta-P, Integrin beta-7, Ly69, M290 IEL Ag
Host Species Rat
Immunogen αIELβ7 integrin from the C57BL/10 mouse T-cell hybridoma MTC-1
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.