Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin α7 (ITGA7) Mouse anti-Mouse, Human, PE, Clone: 3C12, BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568719
This item is not returnable.
View return policy
Description
Integrin α7 (ITGA7) Monoclonal antibody specifically detects Integrin α7 (ITGA7) in Mouse, Human samples. It is validated for Flow cytometry (Routinely Tested)
Specifications
Integrin α7 (ITGA7) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Mouse, Human | |
Purified |
Flow Cytometry | |
3C12 | |
PE | |
ITGA7; Integrin α7; integrin alpha-7; α7 Integrin | |
Mouse Myoblasts | |
0.1 mg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Safety and Handling
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction