Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin Alp v Bta 5 Mouse, anti-Human, R718, Clone: ALULA, BD OptiBuild™

Catalog No. BDB751738 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB751738 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB751738 Supplier BD Biosciences Supplier No. 751738
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The ALULA monoclonal antibody specifically binds to Integrin alpha v beta 5 (αvβ5), which is also known as CD51/β5. Integrin αvβ5 is widely expressed by fibroblasts, endothelial cells, and vascular smooth muscle cells. It is involved in activation-dependent cell migration and cell adhesion to the extracellular matrix. Integrin αvβ5 plays a major role in growth factor-induced angiogenesis in response to VEGF and TGF. The αv Integrins, αvβ5 and αvβ3, αre involved in tumor growth and are being targeted in cancer therapies. The ALULA clone specifically recognizes αvβ5 integrin and functionally blocks the interaction between αvβ5 and vitronectin. Although raised against mouse αvβ5, the ALULA antibody crossreacts with human αvβ5.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin Alp v Bta 5
Applications Flow Cytometry
Classification Monoclonal
Clone ALULA
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.