Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 1/CD49a Antibody (CL7207), Novus Biologicals™
SDP

Catalog No. p-200080826 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP276478 100 μL
NB392759 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP276478 Supplier Novus Biologicals Supplier No. NBP276478
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

Integrin alpha 1/CD49a Monoclonal specifically detects Integrin alpha 1/CD49a in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Knockdown Validated, Block/Neutralize.

Specifications

Antigen Integrin alpha 1/CD49a
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), KnockDown, Neutralization
Classification Monoclonal
Clone CL7207
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500, Knockdown Validated, Block/Neutralize
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide
Gene Alias CD49 antigen-like family member A, CD49a, CD49a antigen, integrin, alpha 1, Laminin and collagen receptor, very late activation protein 1, VLA-1, VLA1integrin alpha-1
Gene Symbols ITGA1
Host Species Mouse
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: RQISRSFFSGTQERKVQRN
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 3672
Test Specificity Specificity of Integrin alpha 1/CD49a antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.