Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 11 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$181.90 - $487.50

Specifications

Antigen Integrin alpha 11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB095649
SDP
View Documents
Novus Biologicals
NBP30382320UL
20 μL
Each for $181.90
Only null left
Add to Cart
 
NB095648
SDP
View Documents
Novus Biologicals
NBP303823100UL
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

Integrin alpha 11 Polyclonal specifically detects Integrin alpha 11 in Human samples. It is validated for Western Blot.
Specifications

Specifications

Integrin alpha 11
Polyclonal
Purified
RUO
Human
HsT18964, integrin alpha-11, integrin, alpha 11
Recombinant fusion protein containing a sequence corresponding to amino acids 940-1141 of human Integrin alpha 11 (NP_001004439.1).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Unconjugated
Rabbit
Signal Transduction
PBS with 50% glycerol, pH7.3.
22801
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.