Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2/CD49b Antibody (16B4) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10
Specifications
Antigen | Integrin alpha 2/CD49b |
---|---|
Clone | 16B4 |
Concentration | 1 mg/mL |
Dilution | Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready |
Applications | Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation |
Description
Integrin alpha 2/CD49b Monoclonal antibody specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, CyTOFSpecifications
Integrin alpha 2/CD49b | |
1 mg/mL | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Cellular Markers, Innate Immunity, Signal Transduction | |
PBS | |
3673 | |
IgG1 | |
Protein A purified |
16B4 | |
Flow Cytometry 1:10 - 1:1000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
Purified human beta 1 preparation from HT1080 fibrosarcoma cell extract. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title