Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2/CD49b Antibody (31H4), PE/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250145PECY7
Description
Integrin alpha 2/CD49b Monoclonal specifically detects Integrin alpha 2/CD49b in Human samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 2/CD49b | |
Monoclonal | |
PE-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
ITGA2 | |
Purified human b1 integrin preparation from HT1080 fibrosarcoma cell extract. | |
0.1 mL | |
Primary | |
It is specific to the alpha 2 subunit by relative expression of antigen on various cell lines by Flow Cytomery, and its recognition of affinity purified alpha 2 beta 1 in Dot Blot. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
31H4 | |
Flow Cytometry | |
alpha-2 subunit, CD49b antigen, integrin, alpha 2 (CD49B, alpha 2 subunit of VLA-2 receptor), VLA 2 | |
Mouse | |
Protein A purified | |
Cellular Markers, Innate Immunity, Signal Transduction | |
3673 | |
Human | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction