Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
                                    Integrin alpha 2b beta 3 Antibody (10E5), mFluor Violet 450 SE, Novus Biologicals™
 
                                    
                                    
                                    
                                    
                                
                            
                            
                            
                            
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281080MFV450
Description
Integrin alpha 2b beta 3 Monoclonal antibody specifically detects Integrin alpha 2b beta 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Integrin alpha 2b beta 3 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 3674 | |
| Store at 4C in the dark. | |
| IgG κ | 
| Western Blot, ELISA, Immunocytochemistry | |
| 10E5 | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| BDPLT16;BDPLT2;CD41;CD41B;GP2B;GPIIb;GT;GT1;GTA;HPA3;Integrin alpha-IIb;ITGA2B;PPP1R93 | |
| Human blood platelets. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction
            