Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2b beta 3, Rabbit anti-Human, Clone: 1000000, Novus Biologicals™

Rabbit Monoclonal Antibody
$268.00 - $584.50
Specifications
Antigen | Integrin alpha 2b beta 3 |
---|---|
Clone | 1000000 |
Concentration | 1 mg/mL |
Dilution | Western Blot, ELISA, Immunocytochemistry/Immunofluorescence |
Applications | Western Blot, ELISA, Immunofluorescence |
Description
Integrin alpha 2b beta 3 Monoclonal specifically detects Integrin alpha 2b beta 3 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Integrin alpha 2b beta 3 | |
1 mg/mL | |
Western Blot, ELISA, Immunofluorescence | |
Unconjugated | |
Rabbit | |
Human | |
3674 | |
Human blood platelets. | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
1000000 | |
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
Monoclonal | |
Purified | |
RUO | |
PBS | |
ITGA2B | |
IgG κ | |
Protein A purified | |
The antibody binds to Integrin alpha 2b beta 3 with an affinity (Kd) of 16.2+-6 nM. The antibody completely inhibits platelet aggregation at 7.8 ug/ml. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title