Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2b/CD41 Antibody (7E3 (Abciximab)), mFluor Violet 450 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281051MFV450
Description
Integrin alpha 2b/CD41 Monoclonal antibody specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Flow CytometrySpecifications
Integrin alpha 2b/CD41 | |
Monoclonal | |
mFluor Violet 450 SE | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry | |
7E3 (Abciximab) | |
Flow Cytometry | |
CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
CD41 is the integrin alpha chain 2b forming a fibrinogen receptor in heterodimeric complex with beta 3. | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
3674 | |
Store at 4C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction