Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2b/CD41 Antibody (IVA30), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB500496PE
Description
Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Human, Bovine samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 2b/CD41 | |
Monoclonal | |
PE | |
CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
The antibody IVA30 reacts with CD41/CD61 complex (bovine), one of the earliest markers of the megakaryocytic lineage. CD41/CD61 complex acts as the receptor for fibrinogen, fibronectin, Von Willebrand factor, vitronectin, thrombin and mediates platelets aggregation. CD41/CD61 is expressed only by platelets and megakaryocytes; the complex may be absent or strongly reduced in Glanzmann's thrombasthenia (GT). | |
Store at 4C in the dark. |
Flow Cytometry | |
IVA30 | |
Flow Cytometry | |
ITGA2B | |
Bovine trombocytes. | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
3674 | |
Human, Bovine | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction