Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2b/CD41 Antibody (M148), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB1002614AF594
Description
Integrin alpha 2b/CD41 Monoclonal antibody specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, ImmunofluorescenceSpecifications
| Integrin alpha 2b/CD41 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
| Homogenized human medulloblastoma tissue. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
| 3674 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence | |
| M148 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction