Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha 2b/CD41 Antibody (MWReg30), FITC, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$390.00 - $780.00

Specifications

Antigen Integrin alpha 2b/CD41
Clone MWReg30
Dilution Flow Cytometry 1:10-1:1000
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14401205
SDP
View Documents
Novus Biologicals
NBP1440120.05MG
0.05 mg
Each for $390.00
Only null left
Add to Cart
 
NBP14401201
SDP
View Documents
Novus Biologicals
NBP1440120.1MG
0.1 mg
Each for $622.00
Only null left
Add to Cart
 
NBP1440125
SDP
View Documents
Novus Biologicals
NBP1440120.5MG
0.5 mg
Each for $780.00
Only null left
Add to Cart
 
Description

Description

Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

Integrin alpha 2b/CD41
Flow Cytometry 1:10-1:1000
Monoclonal
Rat
Mouse
CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK
ITGA2B
IgG1 κ
Affinity Purified
MWReg30
Flow Cytometry
FITC
RUO
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
3674
The immunogen for this antibody was CD41.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.