Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2b/CD41 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $646.00
Specifications
Antigen | Integrin alpha 2b/CD41 |
---|---|
Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Integrin alpha 2b/CD41 Polyclonal specifically detects Integrin alpha 2b/CD41 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Integrin alpha 2b/CD41 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
ITGA2B | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Human | |
P08514 | |
3674 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:CVPQLQLTASVTGSPLLVGADNVLELQMDAANEGEGAYEAELAVHLPQGAHYMRALSNVEGFERLICNQKKENETRV | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title