Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 2b/CD41 Antibody (PM6/248), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10063779AF405
Description
Integrin alpha 2b/CD41 Monoclonal specifically detects Integrin alpha 2b/CD41 in Human, Mustelid, Baboon samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 2b/CD41 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
CD41, CD41 antigen, CD41BHPA3, GP2Bintegrin alpha-IIb, GPalpha IIb, GPIIb, GTA, integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41), integrin, alpha 2b (platelet glycoprotein IIb of IIb/IIIa complex, antigenCD41B), ITGAB, platelet fibrinogen receptor, alpha subunit, Platelet membrane glycoprotein IIb, platelet-specific antigen BAK | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB100-63779 recognizes the human CD41 cell surface antigen, a 140kD glycoprotein expressed by platelets and megakaryocytes. CD41 is also known as platelet glycoprotein IIb, and functions as a receptor for fibrinogen, fibronectin and vWF. It has not been established if clone PM6/248 recognizes free CD41 or CD41 only when complexed with CD61. However, antibody binding is reduced in the presence of EDTA suggesting that the epitope recognized is dependent upon an intact CD41/61 complex. | |
Store at 4C in the dark. | |
IgG1 |
Flow Cytometry | |
PM6/248 | |
Flow Cytometry | |
P08514 | |
ITGA2B | |
Human platelet plasma membrane | |
0.1 mL | |
Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Extracellular Matrix, Immunology, Signal Transduction, Stem Cell Markers | |
3674 | |
Human, Mustelid, Baboon | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction