Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4 beta 1 Antibody (Hu114), PE/Atto594, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals FAB10603PEATT594
Description
Integrin alpha 4 beta 1 Monoclonal antibody specifically detects Integrin alpha 4 beta 1 in Human samples. It is validated for Flow CytometrySpecifications
Integrin alpha 4 beta 1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS | |
Human | |
Protein A or G purified from cell culture supernatant | |
RUO | |
3676 | |
Store at 4C in the dark. Do not freeze. | |
IgG1 |
Flow Cytometry | |
Hu114 | |
Flow Cytometry | |
CD49d, IA4 | |
Human Integrin alpha 4 beta 1 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction