Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4 beta 7/LPAM-1 Antibody (DATK32), HRP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP307899H
Description
Integrin alpha 4 beta 7/LPAM-1 Monoclonal specifically detects Integrin alpha 4 beta 7/LPAM-1 in Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Integrin alpha 4 beta 7/LPAM-1 | |
Monoclonal | |
HRP | |
PBS | |
Rat | |
Protein A purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Immunohistochemistry, Immunohistochemistry (Frozen) | |
DATK32 | |
Immunohistochemistry, Immunohistochemistry-Frozen | |
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
TK1 cells | |
0.1 mL | |
Innate Immunity | |
3676 | |
Store at 4C in the dark. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction