Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4 beta 7/LPAM-1 Antibody (DATK32), PE/Atto594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143283PEATT594
Description
Integrin alpha 4 beta 7/LPAM-1 Monoclonal specifically detects Integrin alpha 4 beta 7/LPAM-1 in Mouse samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 4 beta 7/LPAM-1 | |
Monoclonal | |
PE/ATTO 594 | |
PBS with 0.05% Sodium Azide | |
ITGA4 | |
The immunogen for this antibody was LPAM1. | |
0.1 mL | |
Primary | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
DATK32 | |
Flow Cytometry | |
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
Rat | |
Protein A or G purified | |
Innate Immunity | |
3676.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction