Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4/CD49d Antibody (21.6), DyLight 350, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP281092UV
Description
Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.Specifications
Integrin alpha 4/CD49d | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate | |
ITGA4 | |
Integrin alpha 4/CD49d | |
0.1 mL | |
Cancer, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Microautophagy, Neuronal Cell Markers, Phospho Specific | |
3676 | |
Human | |
Purified |
ELISA, Immunocytochemistry, Immunofluorescence | |
21.6 | |
ELISA, Immunocytochemistry/Immunofluorescence | |
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
This antibody binds to human Integrin alpha 4/CD49d | |
Store at 4°C in the dark. | |
IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction