Learn More
Integrin alpha 4/CD49d Antibody (9F10) - BSA Free, Novus Biologicals™
SDP
Mouse Monoclonal Antibody
Supplier:  Novus Biologicals NBP1433980.1MG
 View more versions of this product
Catalog No. NBP14339801
Only null left
Add to Cart
Description
Description
Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Immunofluorescence.
Specifications
Specifications
Integrin alpha 4/CD49d
Monoclonal
0.5 mg/mL
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Functional
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
Mouse
Affinity Purified
RUO
3676
Store at 4C. Do not freeze.
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Functional Assay
9F10
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
ITGA4
The immunogen for this antibody was CD49d.
0.1 mg
Primary
Human
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only