Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4/CD49d Antibody (HP2/1), PE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB12022858PE
Description
Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 4/CD49d | |
Monoclonal | |
PE | |
PBS with 0.05% Sodium Azide | |
ITGA4 | |
JM leukaemia line (Human). | |
0.1 mL | |
Cancer, Cellular Markers, Cytoskeleton Markers, Phospho Specific | |
3676 | |
Human | |
Purified |
Flow Cytometry | |
HP2/1 | |
Flow Cytometry | |
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse anti Human CD49d monoclonal antibody, clone HP2/1 recognizes human CD49d also known as integrin alpha-4 or VLA-4 subunit alpha. CD49d is a ∼150kDa single pass type 1 transmembrane glycoprotein with seven FG-GAP repeats, characteristic of alpha integrins, in its extracellular domain. CD49d can be proteolytically cleaved to yield framents of 80 and 70kDa (Hemler et al. 1987). CD49d associates with either CD29 to form VLA-4 or with Integrin beta-7 to form The Peyer patches-specific homing receptor LPAM-1, involved in the lymphocyte migration and homing to gut-associated lymphoid tissue (Sackstein 2006) through its interaction with MadCam-1, preferentially expressed on Peyer's patch high endothelial venules and postcapillary venules in lamina propria (Briskin et al. 1997). Mouse anti human CD49d, clone HP2/1 binds to both intact and the 80kDa fragment of integrin alpha-4. CD49d is expressed on monocytes, T cells, B cells, thymocytes and Langerhans cells (de Graaf et al. 1995). Mouse anti Human CD49d, clone HP2/1 can be used in basic studies of VLA-4 mediated adhesion and its interaction with the VCAM-1 structure and has been demonstrated to inhibit cell binding to soluble VCAM-1 (Weller et al. 1991). | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction