Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4/CD49d Antibody (Natalizumab), PE, Novus Biologicals™

Human Monoclonal Antibody
Supplier: Novus Biologicals NBP275916PE
Description
Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for Flow Cytometry.Specifications
| Integrin alpha 4/CD49d | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| ITGA4 | |
| The RAMOS cell line, a non-EBV releasing cell line, injected into mice. | |
| 0.1 mL | |
| Primary | |
| This antibody binds to the alpha 4 subunit of (alpha)4(beta)1 (also known as very late antigen 4 (VLA-4) or CD49d-CD29) and (alpha)4(beta)7 integrin | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| Natalizumab | |
| Flow Cytometry | |
| 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
| Human | |
| Protein A purified | |
| Cancer, Cellular Markers, Cytoskeleton Markers, Phospho Specific | |
| 3676.0 | |
| Human | |
| IgG4 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction