Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha-4 (CD49d) Mouse anti-Rat, Clone: TA-2, MilliporeSigma™

Catalog No. MABF950MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABF950MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABF950MI Supplier MilliporeSigma Supplier No. MABF950
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This mouse monoclonal Anti-Integrin alpha-4 (CD49d), clone TA-2 Immunoprecipitation, and in functional studies as Agonist or Inhibitor for the detection of CD49d/Integrin alpha-4.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Integrin alpha-4 (CD49d)
Applications Inhibition Assays, Flow Cytometry, Immunoprecipitation
Classification Monoclonal
Clone TA-2
Concentration Please refer to lot specific datasheet.
Conjugate Unconjugated
Gene Symbols Itga4
Host Species Mouse
Immunogen Small peritoneal exudate lymphocytes (sPELs).
Purification Method Protein G Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cell Structure
Primary or Secondary Primary
Test Specificity Clone TA-2 detects CD49d/Integrin alpha-4 in Rat splenocytes and lymphocytes.
Target Species Rat
Content And Storage Stable for 1 year at -20°C from date of receipt.Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.