Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 4/CD49d, Rabbit anti-Human, Clone: 21.6, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP2810920.2MG
Description
Integrin alpha 4/CD49d Monoclonal specifically detects Integrin alpha 4/CD49d in Human samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Block/Neutralize.Specifications
| Integrin alpha 4/CD49d | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA, Block/Neutralize | |
| 269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody binds to human Integrin alpha 4/CD49d | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| ELISA, Blocking Assay | |
| 21.6 | |
| Unconjugated | |
| PBS | |
| ITGA4 | |
| Integrin alpha 4/CD49d | |
| 0.2 mg | |
| Cancer, Cell Biology, Cellular Markers, Cytokine Research, Growth and Development, Lysosome Markers, Mast Cell Markers, Membrane Trafficking and Chaperones, Microautophagy, Neuronal Cell Markers, Phospho Specific | |
| 3676 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction