Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 5 beta 1 Antibody (M200 (Volociximab)), Alexa Fluor™ 350 - Chimeric, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252680AF350
Description
Integrin alpha 5 beta 1 Monoclonal specifically detects Integrin alpha 5 beta 1 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence.Specifications
Integrin alpha 5 beta 1 | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
ITGA5 | |
Heterodimeric alpha 5 Beta 1 Fc fusion protein | |
0.1 mL | |
3678 | |
Human | |
IgG κ |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
M200 (Volociximab) | |
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunofluorescence | |
CD49 antigen-like family member E, CD49e, Fibronectin receptor subunit alpha, fibronectin receptor, alpha polypeptide, fibronectin receptor, alpha subunit, FNRA, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), ITGA5/SLC9B1 fusion, very late activation protein 5, alpha subunit, VLA-5, VLA5A | |
Rabbit | |
Protein A or G purified | |
Primary | |
The antibody binds to human (alpha)5(beta)1 integrin with a Kd of 0.367 nM (determined by Biacore), an EC50 of 0.2 nM and an IC50 of 2.3 nM. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction