Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 5/CD49e Antibody (SNAKA52), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250147B
Description
Integrin alpha 5/CD49e Monoclonal specifically detects Integrin alpha 5/CD49e in Human samples. It is validated for ELISA.Specifications
Integrin alpha 5/CD49e | |
Monoclonal | |
Biotin | |
PBS with 0.05% Sodium Azide | |
ITGA5 | |
alpha5 beta1 purified from human placenta. | |
0.1 mL | |
Primary | |
The binding of SNAKA52 is inhibited by anti-beta1 mAbs, indicating that its epitope is close to the interface between the alpha and beta subunits. | |
Store at 4C in the dark. | |
IgG |
ELISA | |
SNAKA52 | |
ELISA | |
CD49 antigen-like family member E, CD49e, CD49e antigen, Fibronectin receptor subunit alpha, fibronectin receptor, alpha subunit, FNRAVLA5A, integrin alpha-5, Integrin alpha-F, integrin, alpha 5 (fibronectin receptor, alpha polypeptide), very late activation protein 5, alpha subunit, VLA-5 | |
Mouse | |
Protein A purified | |
RUO | |
3678 | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction