Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 6 beta 1 Antibody (2548B) [Janelia Fluor™ 549], Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB7809I
Description
Integrin alpha 6 beta 1 Monoclonal antibody specifically detects Integrin alpha 6 beta 1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
Integrin alpha 6 beta 1 | |
Monoclonal | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3655 | |
Store at 4°C in the dark. | |
IgG |
Flow Cytometry, CyTOF | |
2548B | |
Flow Cytometry, CyTOF-ready | |
CD49f, ITGA6B, VLA-6 | |
Chinese Hamster Ovary cell line, CHO-derived Human Integrin alpha 6 beta 1 heterodimer, Phe24-Ser1012(Integrin alpha 6) and Gln21-Asp728 (Integrin beta1), Accession # NP_000201 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction