Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 6/CD49f Antibody (CL6957), Novus Biologicals™

Mouse Monoclonal Antibody
$382.00 - $610.00
Specifications
Antigen | Integrin alpha 6/CD49f |
---|---|
Clone | CL6957 |
Dilution | Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Monoclonal |
Description
Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Integrin alpha 6/CD49f | |
Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Monoclonal | |
Mouse | |
PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
3655 | |
This antibody was developed against Recombinant Protein corresponding to amino acids: APYDDLGKVFIYHGSANGINTKPTQVLKGISPYFGYSIAGNMDLDRNSYPDVAVGSLSDSVTIFRSRPVINIQKTITVTPNRIDLRQKTACGAPSGICLQVKSCFEYTANPAGYNPSISIVGTLEAEKERRKSGLSS | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
CL6957 | |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Human | |
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
ITGA6 | |
IgG1 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title