Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 6/CD49f Antibody (GoH3), DyLight 405, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143400V
Description
Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Integrin alpha 6/CD49f | |
Monoclonal | |
DyLight 405 | |
P23229 | |
ITGA6 | |
The immunogen for this antibody was CD49f. | |
0.1 mL | |
Signal Transduction | |
3655 | |
Human, Mouse, Bovine | |
IgG2a κ |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
GoH3 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
May also react with Bovine. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction