Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 6/CD49f Antibody (GoH3), PE/Cy5.5, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP143400PECY55
Description
Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 6/CD49f | |
Monoclonal | |
PE-Cyanine5.5 | |
PBS with 0.05% Sodium Azide | |
ITGA6 | |
The immunogen for this antibody was CD49f. | |
0.1 mL | |
Primary | |
Human, Mouse, Bovine | |
IgG2a κ |
Flow Cytometry | |
GoH3 | |
Flow Cytometry | |
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
Rat | |
Protein A or G purified | |
Signal Transduction | |
3655.0 | |
Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction