Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 6/CD49f Rabbit anti-Human, Mouse, Rat, Clone: 2R5F1, Novus Biologicals™

Rabbit Monoclonal Antibody
$205.50 - $492.50
Specifications
Antigen | Integrin alpha 6/CD49f |
---|---|
Clone | 2R5F1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Description
Integrin alpha 6/CD49f Monoclonal antibody specifically detects Integrin alpha 6/CD49f in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
Integrin alpha 6/CD49f | |
Western Blot | |
Unconjugated | |
Rabbit | |
Signal Transduction | |
PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
3655 | |
IgG | |
Affinity purified |
2R5F1 | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse, Rat | |
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6B, VLA-6 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 481-591 of human Integrin alpha 6/CD49f (NP_001073286.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title