Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha 8 Antibody (481709), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB6194APCCY7
Description
Integrin alpha 8 Monoclonal antibody specifically detects Integrin alpha 8 in Human samples. It is validated for Flow CytometrySpecifications
Integrin alpha 8 | |
Monoclonal | |
APC-Cyanine7 | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
8516 | |
Store at 4C in the dark. Do not freeze. | |
IgG2b |
Flow Cytometry | |
481709 | |
Flow Cytometry | |
integrin alpha-8, integrin, alpha 8 | |
E. coli-derived recombinant human Integrin alpha 8, Glu610-Leu800, Accession # P53708 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction