Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha D/CD11d Antibody (CA11.8H2) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$206.50 - $499.50
Specifications
Antigen | Integrin alpha D/CD11d |
---|---|
Clone | CA11.8H2 |
Concentration | 1.0 mg/mL |
Dilution | Flow Cytometry 1:10-1:20, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 |
Applications | Immunohistochemistry (Frozen), Immunohistochemistry, Immunoprecipitation, Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017703
![]() |
Novus Biologicals
NB100652920.025MG |
0.025 mg |
Each for $206.50
|
|
|||||
NB10065292H
![]() |
Novus Biologicals
NB100652920.1MG |
0.1 mg |
Each for $499.50
|
|
|||||
Description
Integrin alpha D/CD11d Monoclonal specifically detects Integrin alpha D/CD11d in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.Specifications
Integrin alpha D/CD11d | |
1.0 mg/mL | |
Immunohistochemistry (Frozen), Immunohistochemistry, Immunoprecipitation, Flow Cytometry | |
Unconjugated | |
Mouse | |
Signal Transduction | |
PBS (pH 7.4) with 0.09% Sodium Azide | |
3681 | |
Affinity purified CD11 antigen from canine spleen | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
CA11.8H2 | |
Flow Cytometry 1:10-1:20, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500 | |
Monoclonal | |
Purified | |
RUO | |
Canine | |
ADB2, CD11 antigen-like family member D, CD11D, FLJ39841, integrin alpha-D, integrin, alpha D, leukointegrin alpha D | |
ITGAD | |
IgG1 | |
Protein A purified | |
NB100-65292 recognizes the canine CD11d cell surface antigen (the integrin ad sub-unit). The ad integrin is a newly recognized integrin chain that is expressed as a heterodimer with the b2 integrin (CD18). CD11d is expressed by a sub-population of CD8 positive T cells, macrophages and by gd T cells in the splenic red pulp. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title