Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha D/CD11d Antibody (CA11.8H2), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065292MFV500
Description
Integrin alpha D/CD11d Monoclonal antibody specifically detects Integrin alpha D/CD11d in Canine samples. It is validated for Flow CytometrySpecifications
Integrin alpha D/CD11d | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Canine | |
Purified |
Flow Cytometry | |
CA11.8H2 | |
Flow Cytometry | |
ADB2, CD11 antigen-like family member D, CD11D, FLJ39841, integrin alpha-D, integrin, alpha D, leukointegrin alpha D | |
Affinity purified CD11 antigen from canine spleen | |
0.1 mL | |
Signal Transduction | |
3681 | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction