Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha D/CD11d Antibody (CA11.8H2), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065292AF350
Description
Integrin alpha D/CD11d Monoclonal specifically detects Integrin alpha D/CD11d in Canine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Integrin alpha D/CD11d | |
Monoclonal | |
Alexa Fluor 350 | |
50 mM sodium borate with 0.05% sodium azide | |
ITGAD | |
Affinity purified CD11 antigen from canine spleen | |
Primary | |
NB100-65292 recognizes the canine CD11d cell surface antigen (the integrin ad sub-unit). The ad integrin is a newly recognized integrin chain that is expressed as a heterodimer with the b2 integrin (CD18). CD11d is expressed by a sub-population of CD8 positive T cells, macrophages and by gd T cells in the splenic red pulp. | |
Store at 4°C in the dark. |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
CA11.8H2 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
ADB2, CD11 antigen-like family member D, CD11D, FLJ39841, integrin alpha-D, integrin, alpha D, leukointegrin alpha D | |
Mouse | |
100 μL | |
3681 | |
Canine | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction