Learn More
Description
Specifications
Specifications
| Antigen | Integrin alpha E/CD103 |
| Applications | Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry |
| Classification | Monoclonal |
| Clone | 2G5.1 |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100 |
| Formulation | PBS, 0.5% Bovine Serum Albumin with 0.09% Sodium Azide |
| Gene Accession No. | P38570 |
| Gene Alias | antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
