Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha E/CD103 Antibody (2G5.1), Novus Biologicals™
SDP

Catalog No. p-7200778 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017700 0.025 mg
NB10065271 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017700 Supplier Novus Biologicals Supplier No. NB100652710.025MG
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.

Specifications

Antigen Integrin alpha E/CD103
Applications Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), Flow Cytometry
Classification Monoclonal
Clone 2G5.1
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry 1:50-1:100, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Frozen 1:50-1:100
Formulation PBS, 0.5% Bovine Serum Albumin with 0.09% Sodium Azide
Gene Accession No. P38570
Gene Alias antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Gene Symbols ITGAE
Host Species Mouse
Immunogen Human Intestinal lymphocyte
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 3682
Test Specificity NB100-65271 recognizes the human CD103 cell surface antigen, also known as the alphaE integrin. This reacts with various subsets of human intra-epithelial lymphocytes:- 40% of lamina propria T-cells; 30% of mesenteric lymphoblasts and some lymphocytes from other mucosa. It recognizes less than 0.1% bone marrow cells and less than 0.7% PBL.
Target Species Human, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.