Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha E/CD103 Antibody, Allophycocyanin, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298926APC
Description
Integrin alpha E/CD103 Polyclonal antibody specifically detects Integrin alpha E/CD103 in Human samples. It is validated for Western Blot, Immunocytochemistry/ ImmunofluorescenceSpecifications
Integrin alpha E/CD103 | |
Polyclonal | |
Western Blot, Immunocytochemistry/ Immunofluorescence | |
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human Integrin alpha E/CD103. | |
0.1 mL | |
Innate Immunity | |
3682 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunocytochemistry | |
APC | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction