Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha E/CD103 Antibody (AP-MAB0828), Alexa Fluor™ 594, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP106674AF594
Description
Integrin alpha E/CD103 Monoclonal antibody specifically detects Integrin alpha E/CD103 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)Specifications
Integrin alpha E/CD103 | |
Monoclonal | |
Alexa Fluor 594 | |
50mM Sodium Borate | |
Rat | |
Protein A or G purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
AP-MAB0828 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
Mouse mammary tumor cells | |
0.1 mL | |
Innate Immunity | |
3682 | |
Store at 4°C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction