Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha E/CD103 Antibody (Ber-ACT8) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
$196.10 - $501.50
Specifications
Antigen | Integrin alpha E/CD103 |
---|---|
Clone | Ber-ACT8 |
Concentration | 1 mg/mL |
Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF |
Classification | Monoclonal |
Description
Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen, Flow (Cell Surface), CyTOF-ready.Specifications
Integrin alpha E/CD103 | |
1 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
Human, Primate | |
3682 | |
HTLV-1 induced human T cell line MAPS16 | |
Primary | |
Store at 4C. Do not freeze. |
Ber-ACT8 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen), CyTOF | |
Unconjugated | |
Mouse | |
Innate Immunity | |
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
ITGAE | |
IgG1 κ | |
Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title