Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha E/CD103 Antibody (BP6) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP2504460.025ML
Description
Integrin alpha E/CD103 Monoclonal antibody specifically detects Integrin alpha E/CD103 in Human samples. It is validated for Immunohistochemistry, ImmunoprecipitationSpecifications
Integrin alpha E/CD103 | |
Monoclonal | |
1 mg/mL | |
Immunohistochemistry 1:10 - 1:500, Immunoprecipitation 1:10 - 1:500 | |
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
Hairy cell spleen lysate | |
0.025 mL | |
Innate Immunity | |
3682 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 κ |
Immunohistochemistry, Immunoprecipitation | |
BP6 | |
Unconjugated | |
PBS | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction