Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha E/CD103 Antibody (LF61), DyLight 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10065272UV
Description
Integrin alpha E/CD103 Monoclonal specifically detects Integrin alpha E/CD103 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.Specifications
Integrin alpha E/CD103 | |
Monoclonal | |
DyLight 350 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
ITGAE | |
Hairy cell leukemia cells | |
0.1 mL | |
Innate Immunity | |
3682 | |
Human | |
Purified |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
LF61 | |
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen | |
Mouse | |
Protein G purified | |
RUO | |
Primary | |
NB100-65272 recognizes the human CD103 cell surface antigen, a glycoprotein expressed by approximately 1% of peripheral blood lymphocytes, activated T lymphocytes and by hairy cell leukaemia cells. The antigen is also expressed by intraepithelial lymphocytes. It has recently been shown to be identical to the alpha E integrin. | |
Store at 4C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction