Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Integrin alpha E/CD103 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.50 - $499.50

Specifications

Antigen Integrin alpha E/CD103
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Form Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093068
SDP
View Documents
Novus Biologicals
NBP30354520UL
20 μL
Each for $206.50
Only null left
Add to Cart
 
NB093067
SDP
View Documents
Novus Biologicals
NBP303545100UL
100 μL
Each for $499.50
Only null left
Add to Cart
 
Description

Description

Integrin alpha E/CD103 Polyclonal specifically detects Integrin alpha E/CD103 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

Integrin alpha E/CD103
Polyclonal
Purified
RUO
Human, Mouse, Rat
antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide, CD103, CD103 antigen, HML-1 antigen, HUMINAE, integrin alpha-E, Integrin alpha-IEL, integrin, alpha E (antigen CD103, human mucosal lymphocyte antigen 1; alphapolypeptide), MGC141996, Mucosal lymphocyte 1 antigen
Recombinant fusion protein containing a sequence corresponding to amino acids 19-200 of human Integrin alpha E/CD103 (NP_002199.3).
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry
Unconjugated
Rabbit
Innate Immunity
PBS with 50% glycerol, pH7.3.
3682
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.