Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha L/CD11a Antibody (CRIS-3), Allophycocyanin/Cy7, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP234599APCCY7
Description
Integrin alpha L/CD11a Monoclonal specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Flow Cytometry.Specifications
Integrin alpha L/CD11a | |
Monoclonal | |
APC-Cyanine7 | |
PBS with 0.05% Sodium Azide | |
ITGAL | |
Stimulated human leukocytes | |
0.1 mL | |
Primary | |
Recognizes a protein of 180kDa, identified as CD11a (Leucocyte Workshop II; Code N202). CD11a complex with the 2 subunit of the integrin family, CD18, to form the cell surface heterodimer, LFA-1 or CD11a/C18 (aLbL). LFA-1 is expressed on all leukocytes including lymphocytes, monocytes, and granulocytes. It is involved in leukocyte adhesion to its ligands including intercellular adhesion molecule-1 (ICAM-1 or CD54), ICAM-2 (CD102), ICAM-3 (CD50) and Telencephalin (TLN) and play a role in most immune/inflammatory responses. | |
Store at 4°C in the dark. Do not freeze. |
Flow Cytometry | |
CRIS-3 | |
Flow Cytometry | |
CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 | |
Mouse | |
Protein A or G purified | |
Adaptive Immunity, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
3683.0 | |
Human | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction