Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Integrin alpha L/CD11a Antibody (M17/4) - Low Endotoxin, Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP309088
Description
Integrin alpha L/CD11a Monoclonal specifically detects Integrin alpha L/CD11a in Human samples. It is validated for Flow Cytometry, Immunoprecipitation, Functional.Specifications
Integrin alpha L/CD11a | |
Monoclonal | |
1 mg/ml | |
Flow Cytometry, Immunoprecipitation, Functional | |
CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 | |
Leukocytes from a pacient suffering from a LGL-type leukaemia. | |
0.1 mg | |
Adaptive Immunity, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Myeloid Cell Markers, Signal Transduction, Stem Cell Markers | |
3683 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG1 |
Flow Cytometry, Immunoprecipitation, Functional Assay | |
MEM-25 | |
Unconjugated | |
PBS, pH 7.4, 0.2 um filter sterilized | |
Mouse | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction